Skip to Content
Merck
All Photos(2)

Documents

Safety Information

794155

Sigma-Aldrich

2-Methoxy-5-carboxyphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BO5
CAS Number:
Molecular Weight:
195.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

powder

mp

207-210 °C (lit.)

InChI

1S/C8H9BO5/c1-14-7-3-2-5(8(10)11)4-6(7)9(12)13/h2-4,12-13H,1H3,(H,10,11)

InChI key

UENFRNNKQBQROW-UHFFFAOYSA-N

General description

2-Methoxy-5-carboxyphenylboronic acid (3-borono-4-methoxybenzoic acid) is a monosubstituted aryl boronic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

794155-BULK:
794155-VAR:
794155-1G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service