Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

77983

Sigma-Aldrich

4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluorononyl azide

≥97.0%

Synonym(s):

1H,1H,2H,2H,3H,3H-Perfluorononyl azide, 3-(Perfluorohexyl)propyl azide, 9-Azido-1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorononane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6F13N3
CAS Number:
Molecular Weight:
403.14
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

liquid

refractive index

n20/D 1.342

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCCN=[N+]=[N-]

InChI

1S/C9H6F13N3/c10-4(11,2-1-3-24-25-23)5(12,13)6(14,15)7(16,17)8(18,19)9(20,21)22/h1-3H2

InChI key

DQEDVGBNEFVPEU-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

77983-5MG:
77983-BULK:
77983-25MG:
77983-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service