Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

779687

Sigma-Aldrich

Ethyl 2-cyano-3,3-bis(methylthio)acrylate

≥98.0% (GC)

Synonym(s):

Ethyl 2-cyano-3,3-bis(methylthio)-2-propenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO2S2
CAS Number:
Molecular Weight:
217.31
Beilstein:
1778721
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

solid

composition

sulfur content, 28.9-30.1%

SMILES string

CCOC(=O)\C(C#N)=C(\SC)SC

InChI

1S/C8H11NO2S2/c1-4-11-7(10)6(5-9)8(12-2)13-3/h4H2,1-3H3

InChI key

POABRARINOCORV-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

JAN Code

779687-BULK:
779687-100G:
779687-10G:
779687-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service