Skip to Content
Merck
All Photos(1)

Documents

Safety Information

778648

Sigma-Aldrich

(RP)-1-[(S)-1-(Di-tert-butylphosphino)ethyl]-2-[(S)-phenylphosphinoyl]ferrocene

≥97%

Synonym(s):

(2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-[(S)-phenylphosphinyl]ferrocene (acc to CAS), JoSPOphos SL-J688-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H36FeOP2
CAS Number:
Molecular Weight:
482.36
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥97%

form

powder

optical purity

ee: ≥99%

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2[P@H](=O)c3ccccc3)P(C(C)(C)C)C(C)(C)C

InChI

1S/C21H31OP2.C5H5.Fe/c1-16(24(20(2,3)4)21(5,6)7)18-14-11-15-19(18)23(22)17-12-9-8-10-13-17;1-2-4-5-3-1;/h8-16,23H,1-7H3;1-5H;/t16-;;/m0../s1

InChI key

ZRFUSACJLNCGTO-SQKCAUCHSA-N

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

778648-VAR:
778648-500MG:
778648-BULK:
778648-100MG:
778648-1G:
778648-5G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral mixed secondary phosphine-oxide-phosphines: high-performing and easily accessible ligands for asymmetric hydrogenation.
Heidi Landert et al.
Angewandte Chemie (International ed. in English), 49(38), 6873-6876 (2010-08-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service