Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

775509

Sigma-Aldrich

5-(Aminomethyl)thiazole dihydrochloride

Synonym(s):

5-Thiazolemethanamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2S · 2HCl
CAS Number:
Molecular Weight:
187.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

solid

mp

215-222 °C

functional group

amine

storage temp.

2-8°C

SMILES string

NCC1=CN=CS1.Cl.Cl

InChI

1S/C4H6N2S.2ClH/c5-1-4-2-6-3-7-4;;/h2-3H,1,5H2;2*1H

InChI key

URVNRCYESACKMV-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

775509-VAR:
775509-250MG:
775509-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service