Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

767891

Sigma-Aldrich

1,3-Dihydroxyacetone-2-13C dimer

99% 13C, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C4H12O6
CAS Number:
Molecular Weight:
182.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99% 13C

Assay

95% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+2

storage temp.

2-8°C

SMILES string

OC[13C]1(O)CO[13C](CO)(O)CO1

InChI

1S/C6H12O6/c7-1-5(9)3-12-6(10,2-8)4-11-5/h7-10H,1-4H2/i5+1,6+1

InChI key

KEQUNHIAUQQPAC-MPOCSFTDSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

767891-BULK:
767891-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service