Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

767646

Sigma-Aldrich

2,6-Ditolylbenzo[1,2-b:4,5-b′]dithiophene

sublimed grade, 95%

Synonym(s):

DT-BDT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H18S2
CAS Number:
Molecular Weight:
370.53
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Assay

95%

form

powder or crystals

mp

427-432 °C

semiconductor properties

P-type (mobility=10−2 cm2/V·s)

SMILES string

Cc1ccc(cc1)-c2cc3cc4sc(cc4cc3s2)-c5ccc(C)cc5

InChI

1S/C24H18S2/c1-15-3-7-17(8-4-15)21-11-19-13-24-20(14-23(19)25-21)12-22(26-24)18-9-5-16(2)6-10-18/h3-14H,1-2H3

InChI key

DWVXIELPJDWPNY-UHFFFAOYSA-N

Application

Organic Field Effect Transistor (OFET) Materials; p-Type Organic Semiconductors; p-Type Small Molecules; sublimed grade materials

Citation

Kuwabara, Hirokazu; Yui, Tatuto; Ikeda, Masaaki; Takimiya, Kazuo.
Organic field effect transistor. PCT Int. Appl. (2009), 41pp. CODEN: PIXXD2 WO 2009038120 A1 20090326

Legal Information

Product of Nippon Kayaku

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

767646-500MG:
767646-BULK:
767646-VAR:
767646-100MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Plexcore® organic conductive inks are electronic grade inks formulated for use in the hole injection layers of OLEDs.

Sublimed materials for organic electronic devices such of OFETs and OTFTs allow the achievement of better electronic properties, and may help increase a device’s lifetime.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service