765031
3,6-Bis(N-carbazolyl)-N-phenylcarbazole
99% (HPLC)
Synonym(s):
9′-Phenyl-9,3′:6′,9′′-ter-9H-carbazole, N-Phenyl-3,6-tricarbazolylcarbazole, BCC-36
About This Item
Recommended Products
Assay
99% (HPLC)
form
solid
mp
281-286 °C
λmax
239 nm in THF
Orbital energy
HOMO 5.29 eV
LUMO 1.90 eV
OLED Device Performance
ITO/PEDOT:PSS/BCC-27: FIr6 (10 wt%)/TAZ/Cs2CO3:Al
- Color: blue
- Max. Luminance: 12300 Cd/m2
- Max. EQE: 4.1 %
- Turn-On Voltage: 5 V
ITO/PEDOT:PSS/BCC-27: FIrpic (10 wt%)/TAZ/Cs2CO3:Al
- Color: blue
- Max. Luminance: 19200 Cd/m2
- Max. EQE: 9.7 %
- Turn-On Voltage: 3.9 V
SMILES string
c1ccc(cc1)-n2c3ccc(cc3c4cc(ccc24)-n5c6ccccc6c7ccccc57)-n8c9ccccc9c%10ccccc8%10
InChI
1S/C42H27N3/c1-2-12-28(13-3-1)43-41-24-22-29(44-37-18-8-4-14-31(37)32-15-5-9-19-38(32)44)26-35(41)36-27-30(23-25-42(36)43)45-39-20-10-6-16-33(39)34-17-7-11-21-40(34)45/h1-27H
InChI key
ZEGYQBGJQMTXKA-UHFFFAOYSA-N
Application
- Device-1: ITO/PEDOT:PSS/BCC-27:FIrpic (10 wt%)/TAZ/Cs2CO3/Al
- Device-2: ITO/PEDOT:PSS/BCC-27:FIr6 (10 wt%)/TAZ/Cs2CO3/Al
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
765031-1G:
765031-BULK:
765031-VAR:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.
Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service