Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

764191

Sigma-Aldrich

Alkyne-PEG5-N-hydroxysuccinimidyl ester

Synonym(s):

Acetylene-PEG5-NHS ester, Alkyne-PEG5-NHS ester, Alkyne-PEG5-succinimidyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
¥22,400

¥22,400


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
¥22,400

About This Item

Empirical Formula (Hill Notation):
C18H27NO9
Molecular Weight:
401.41
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

¥22,400


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

functional group

NHS ester
alkyne

storage temp.

−20°C

SMILES string

C#CCOCCOCCOCCOCCOCCC(ON1C(CCC1=O)=O)=O

InChI

1S/C18H27NO9/c1-2-6-23-8-10-25-12-14-27-15-13-26-11-9-24-7-5-18(22)28-19-16(20)3-4-17(19)21/h1H,3-15H2

InChI key

ACZDUKRWTPZVRP-UHFFFAOYSA-N

Application

Heterobifunctional crosslinker enabling the connection of an azide containing compound or biomolecule to one with a primary amine. The succinimidyl ester group will react preferentially with primary amines to form a covalent adduct and the terminal alkyne will react with azides via copper-catalyzed 1,3-dipolar cycloaddition click chemistry. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification amine containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

764191-BULK:
764191-25MG:
764191-VAR:
764191-10MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service