Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

758272

Sigma-Aldrich

8-Ethylquinoline N-oxide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO
Molecular Weight:
173.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: oxidant

mp

104-109 °C

storage temp.

2-8°C

SMILES string

CCC1=CC=CC2=C1[N+]([O-])=CC=C2

InChI

1S/C11H11NO/c1-2-9-5-3-6-10-7-4-8-12(13)11(9)10/h3-8H,2H2,1H3

InChI key

LZEBWZDDNNWJJK-UHFFFAOYSA-N

Application

Reactant for the preparation of:
  • α, β-unsaturated enones with catalytic amounts of Gold complexes
  • Synthesis of azetidines (4-membered nitrogen containing heterocycles) through an intramolecular oxidative cyclization in the presence of catalytic amounts of Gold complexes

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

758272-VAR:
758272-5G:
758272-1G:
758272-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A flexible and stereoselective synthesis of azetidin-3-ones through gold-catalyzed intermolecular oxidation of alkynes.
Longwu Ye et al.
Angewandte Chemie (International ed. in English), 50(14), 3236-3239 (2011-03-08)
Biao Lu et al.
Journal of the American Chemical Society, 132(40), 14070-14072 (2010-09-22)
Gold-catalyzed intermolecular oxidations of internal alkynes have been achieved with high regioselectivities using 8-alkylquinoline N-oxides as oxidants and in the absence of acid additives. Synthetically versatile α,β-unsaturated carbonyls are obtained in good to excellent yields and with excellent E-selectivities. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service