Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

756709

Sigma-Aldrich

Bisphenol A acetate propionate

technical grade

Synonym(s):

EC-326, Epoxy Curative

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

97% (as a mixture)

form

solid

impurities

~25% BPA diacetate
~25% BPA dipropionate

viscosity

2,000 cP(25 °C)

mp

39-44 °C

SMILES string

CCC(=O)Oc1ccc(cc1)C(C)(C)c2ccc(OC(C)=O)cc2

InChI

1S/C20H22O4/c1-5-19(22)24-18-12-8-16(9-13-18)20(3,4)15-6-10-17(11-7-15)23-14(2)21/h6-13H,5H2,1-4H3

InChI key

VWMUCEDXVAQLNS-UHFFFAOYSA-N

General description

This contains a mixture of three functionalized bisphenol A curing agents: diacetate, propionate/acetate, and dipropionate. The overall concentration of these three curing agents is >97%

Application

Propanioc acid, mixed diesters with acetic acid and bisphenol A
  • Hydrolytically resistant
  • Low melting point
  • Thermal stability
  • Hydrophobic
  • Toughener
  • Does not impede free radical cure
  • Well-suited for film adhesives and pre-applied adhesives

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

756709-VAR:
756709-BULK:
756709-25G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service