Skip to Content
Merck
All Photos(4)

Key Documents

Safety Information

748730

Sigma-Aldrich

Methyl 1H-imidazole-1-carboxylate

95%

Synonym(s):

Heller-Sarpong Reagent, N-Carbomethoxyimidazole, MImC, Methylimidazolecarbamate, Sarpong reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

38-44 °C

storage temp.

−20°C

SMILES string

COC(=O)n1ccnc1

InChI

1S/C5H6N2O2/c1-9-5(8)7-3-2-6-4-7/h2-4H,1H3

InChI key

JQCPMLVMNQVRCC-UHFFFAOYSA-N

General description

Methyl 1H-imidazole-1-carboxylate is a reagent developed by Heller and Sarpong for the amidation and esterification of carboxylic acids chemoselectively.

Application

Methyl 1H-imidazole-1-carboxylate can be used to prepare fluorous β-keto esters, precursors used for the synthesis of fluorinated L-carbidopa derivatives.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

194.0 °F

Flash Point(C)

90 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

748730-25G:
748730-BULK:
748730-5G:
748730-VAR:
748730-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorous l-Carbidopa Precursors: Highly Enantioselective Synthesis and Computational Prediction of Bioactivity
Granados A, et al.
The Journal of Organic Chemistry, 83(1), 303-313 (2018)
Chemoselective esterification and amidation of carboxylic acids with imidazole carbamates and ureas
Heller ST and Sarpong R
Organic Letters, 12(20), 4572-4575 (2010)

Related Content

Professor Heller's research focuses on mild acylation reactions using carbonylimidazole-derived reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service