Skip to Content
Merck
All Photos(1)

Documents

Safety Information

748641

Sigma-Aldrich

Furan-3-boronic acid MIDA ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10BNO5
Molecular Weight:
222.99
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

solid

mp

132-134 °C

storage temp.

2-8°C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2ccoc2

InChI

1S/C9H10BNO5/c1-11-4-8(12)15-10(16-9(13)5-11)7-2-3-14-6-7/h2-3,6H,4-5H2,1H3

InChI key

FKZKTGSSMZITRF-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

748641-VAR:
748641-5G:
748641-BULK:
748641-1G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service