Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

746592

Sigma-Aldrich

Heptanal-d14

≥98 atom % D, ≥96% (CP)

Synonym(s):

Deuterated Heptanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)5COD
Molecular Weight:
128.27
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥98 atom % D

Assay

≥96% (CP)

form

liquid

mass shift

M+14

SMILES string

O=C(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H]

InChI

1S/C7H14O/c1-2-3-4-5-6-7-8/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D

InChI key

FXHGMKSSBGDXIY-OBYKGMMLSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

746592-VAR:
746592-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service