Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

744980

Sigma-Aldrich

1-(Azidomethyl)-2-methylbenzene solution

~0.5 M in tert-butyl methyl ether, ≥97.0% (HPLC)

Synonym(s):

α-Azido-o-xylene solution, o-Methylbenzyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N3
CAS Number:
Molecular Weight:
147.18
Beilstein:
4860454
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

liquid

reaction suitability

reaction type: click chemistry

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

2-8°C

SMILES string

Cc1ccccc1CN=[N+]=[N-]

InChI

1S/C8H9N3/c1-7-4-2-3-5-8(7)6-10-11-9/h2-5H,6H2,1H3

InChI key

TWZXVFYNRLIXRY-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-18.4 °F

Flash Point(C)

-28 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

744980-BULK:
744980-50ML:4548174012185
744980-VAR:
744980-10ML:4548174012178


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meina Liu et al.
Organic letters, 13(5), 1102-1105 (2011-02-12)
A structurally well-defined copper(I) isonitrile complex is shown to be an efficient, heterogeneous catalyst for the Huisgen azide-alkyne 1,3-dipolar cycloaddition under mild conditions in water. Notably, this catalyst can also be utilized in a three-component reaction of halides, sodium azide
Atif Sarwar et al.
PloS one, 10(4), e0123084-e0123084 (2015-05-01)
Recently, the attention of researchers has been drawn toward the synthesis of chitosan derivatives and their nanoparticles with enhanced antimicrobial activities. In this study, chitosan derivatives with different azides and alkyne groups were synthesized using click chemistry, and these were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service