Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

742961

Sigma-Aldrich

4-Methoxybenzyl bromide

≥90% (GC)

Synonym(s):

1-(Bromomethyl)-4-methoxybenzene, 4-(Bromomethyl)anisole, p-(Bromomethyl)anisole, p-Methoxybenzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2BrC6H4OCH3
CAS Number:
Molecular Weight:
201.06
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

≥90% (GC)

contains

~3 wt. % K2CO3 as stabilizer

refractive index

n20/D 1.573
n20/D 1.5780 (lit.)

bp

91 °C/1 mmHg (lit.)

density

1.395 g/mL at 19 °C (lit.)
1.432 g/mL at 20 °C

SMILES string

COc1ccc(CBr)cc1

InChI

1S/C8H9BrO/c1-10-8-4-2-7(6-9)3-5-8/h2-5H,6H2,1H3

InChI key

GIGRWGTZFONRKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

742961-1ML:
742961-5ML:
742961-VAR:
742961-25ML:
742961-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric total syntheses of (R)-(-)-argentilactone and (S)-5-hexadecanolide.
Sabitha, G., et al.
Synthesis, 17, 2879-2884 (2006)
Kelvine Chignen Possi et al.
Journal of medicinal chemistry, 60(22), 9263-9274 (2017-10-14)
Azapeptide analogues of growth hormone releasing peptide-6 (GHRP-6) exhibit promising affinity, selectivity, and modulator activity on the cluster of differentiation 36 receptor (CD36). For example, [A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service