Skip to Content
Merck
All Photos(1)

Documents

Safety Information

74231

Sigma-Aldrich

Tris[3,5-bis(trifluoromethyl)phenyl]phosphine

≥95.0% (GC), yellow-brown

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H9F18P
CAS Number:
Molecular Weight:
670.27
Beilstein:
6556841
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

color

yellow-brown

functional group

phosphine

SMILES string

FC(F)(F)c1cc(cc(c1)C(F)(F)F)P(c2cc(cc(c2)C(F)(F)F)C(F)(F)F)c3cc(cc(c3)C(F)(F)F)C(F)(F)F

InChI

1S/C24H9F18P/c25-19(26,27)10-1-11(20(28,29)30)5-16(4-10)43(17-6-12(21(31,32)33)2-13(7-17)22(34,35)36)18-8-14(23(37,38)39)3-15(9-18)24(40,41)42/h1-9H

InChI key

ITJHLZVYLDBFOJ-UHFFFAOYSA-N

Other Notes

Phosphine with enhanced solubility in solvents

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

74231-500MG-F:
74231-BULK-F:
74231-VAR-F:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.K. Morita et al.
Chemical Communications (Cambridge, England), 1397-1397 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service