Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

733997

Sigma-Aldrich

2-Bromo-4-(difluoromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
¥17,300

¥17,300


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
¥17,300

About This Item

Empirical Formula (Hill Notation):
C6H4BrF2N
CAS Number:
Molecular Weight:
208.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

¥17,300


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.520

density

1.657 g/mL at 25 °C

functional group

bromo
fluoro

storage temp.

−20°C

SMILES string

FC(F)c1ccnc(Br)c1

InChI

1S/C6H4BrF2N/c7-5-3-4(6(8)9)1-2-10-5/h1-3,6H

InChI key

IAFLELWFIHZCGT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 1

Flash Point(F)

217.9 °F

Flash Point(C)

103.3 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

733997-BULK:
733997-VAR:
733997-500MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service