Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

733695

Sigma-Aldrich

1,1,1-Trifluoro-2-(trifluoromethyl)-2-hydroxy-4-methyl-5-pentyl methacrylate

>97%

Synonym(s):

5,5,5-Trifluoro-4-hydroxy-2-methyl-4-(trifluoromethyl)pentyl methacrylate, HFA monomer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14F6O3
CAS Number:
Molecular Weight:
308.22
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

>97%

form

liquid

density

1.302 g/mL at 25 °C

SMILES string

CC(COC(=O)C(C)=C)CC(O)(C(F)(F)F)C(F)(F)F

InChI

1S/C11H14F6O3/c1-6(2)8(18)20-5-7(3)4-9(19,10(12,13)14)11(15,16)17/h7,19H,1,4-5H2,2-3H3

InChI key

WNAVFWSIXYHORQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

733695-1G:
733695-VAR:
733695-BULK:
733695-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service