Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

724165

Sigma-Aldrich

Ethyl 1-methylpiperidine-4-carboxylate

97%

Synonym(s):

1-Methylpiperidine-4-carboxylic acid ethyl ester, Ethyl N-methylisonipecotate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.450

density

0.963 g/mL at 25 °C

functional group

ester

SMILES string

CCOC(=O)C1CCN(C)CC1

InChI

1S/C9H17NO2/c1-3-12-9(11)8-4-6-10(2)7-5-8/h8H,3-7H2,1-2H3

InChI key

JWXOOQCMGJBSML-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Piperidine derivatives as inhibitors of S. aureus and E. coli enoyl-ACP reductase
  • GABAA receptor agonists
  • Water-soluble DNA-binding subunits for analogues of cytotoxic antibiotic CC-1065 and their prodrugs
  • Tricyclic pharmaceuticals
  • 5-HT3 antagonists

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.0 °F - closed cup

Flash Point(C)

87.8 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

724165-BULK:
724165-1G:
724165-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service