71941
4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoyl chloride
≥97.0% (GC)
Synonym(s):
2H,2H,3H,3H-Perfluoroundecanoyl chloride
About This Item
Recommended Products
Quality Level
Assay
≥97.0% (GC)
form
crystals
SMILES string
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCC(Cl)=O
InChI
1S/C11H4ClF17O/c12-3(30)1-2-4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)9(23,24)10(25,26)11(27,28)29/h1-2H2
InChI key
QYLZAPXXSYDKKB-UHFFFAOYSA-N
Physical form
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1
Target Organs
Liver
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
71941-1G-F:
71941-5G-F:
71941-BULK-F:
71941-VAR-F:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service