Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

718327

Sigma-Aldrich

Ethyl 5-[(4-methylphenyl)sulfonyl]-3-oxopentanoate

95%

Synonym(s):

5-[(4-Methylphenyl)sulfonyl]-3-oxopentanoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O5S
CAS Number:
Molecular Weight:
298.35
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

42-46 °C

functional group

ester
ketone
sulfone

SMILES string

CCOC(=O)CC(=O)CCS(=O)(=O)c1ccc(C)cc1

InChI

1S/C14H18O5S/c1-3-19-14(16)10-12(15)8-9-20(17,18)13-6-4-11(2)5-7-13/h4-7H,3,8-10H2,1-2H3

InChI key

APRUPJUUTCSBAE-UHFFFAOYSA-N

Application

Ethyl 5-[(4-methylphenyl)sulfonyl]-3-oxopentanoate can be used as a reactant to prepare:
  • Ethyl 3-oxopent-4-enoate (Nazarov′s reagent) via base-induced β-elimination reaction. Nazarov′s reagent can be employed as an anulating agent in Robinson annulation of cyclic β-diketones and cycloalkanones.
  • γ-pyrones via triflic anhydride-mediated electrophilic condensation reaction.

Reactant for:
  • Preparation of the Nazarov reagent via base-induced ß-elimination reaction

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

718327-1G:
718327-BULK:
718327-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethyl 5-[(4-Methylphenyl) sulfonyl]-3-Oxopentanoate: A Bench-Stable Synthon for Ethyl 3-Oxopent-4-enoate (Nazarov?s Reagent)
Benetti S, et al.
Synlett, 2008(17), 2609-2612 (2008)
Direct synthesis of γ-pyrones by electrophilic condensation of β-ketoesters
Rodrigues CAB, et al.
Organic & Biomolecular Chemistry, 15(3), 680-683 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service