Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

717371

Sigma-Aldrich

(R)-1-[(Sp)-2-(Diphenylphosphino)ferrocenyl]ethylamine

97%

Synonym(s):

(Sp)-1-[(1R)-(1-Aminoethyl)]-2-(diphenylphosphino)ferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24FeNP
Molecular Weight:
413.27
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

131-135 °C

functional group

amine
phosphine

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H](N)[C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4

InChI

1S/C19H19NP.C5H5.Fe/c1-15(20)18-13-8-14-19(18)21(16-9-4-2-5-10-16)17-11-6-3-7-12-17;1-2-4-5-3-1;/h2-15H,20H2,1H3;1-5H;/t15-;;/m1../s1

InChI key

DRBVPPSGNFMTKZ-QCUBGVIVSA-N

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

717371-BULK:
717371-VAR:
717371-100MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service