Skip to Content
Merck
All Photos(1)

Documents

Safety Information

711020

Sigma-Aldrich

Cytidine-13C9 5′-triphosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom % 13C, ≥95% (CP)

Synonym(s):

CTP-13C9 disodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C9H14N3Na2O14P3
Molecular Weight:
536.05
UNSPSC Code:
41106305
NACRES:
NA.12

isotopic purity

≥98 atom % 13C

Assay

≥95% (CP)

form

solution

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+9

shipped in

dry ice

storage temp.

−20°C

SMILES string

N[13C]([13CH]=[13CH]N1[13C@H](O[13C@@H]2[13CH2]OP(OP(OP(O)(O[Na])=O)(O[Na])=O)(O)=O)[13C@H](O)[13C@@H]2O)=N[13C]1=O

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

Molecular weight and empirical formula based on the free acid.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

711020-BULK:
711020-10MG-PW:
711020-1MG-PW:
711020-1MG:
711020-10MG:
711020-25MG-PW:
711020-25MG:
711020-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service