Skip to Content
Merck
All Photos(1)

Documents

Safety Information

709387

Sigma-Aldrich

Cyanamide-13C,15N2

98 atom % 15N, 99 atom % 13C, 98% (CP), anhydrous (stabilized with 0.1% acetic acid)

Synonym(s):

Carbodiimide-13C,15N2, Hydrogen cyanamide-13C,15N2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H215N13C15N
CAS Number:
Molecular Weight:
45.02
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

grade

anhydrous (stabilized with 0.1% acetic acid)

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

98% (CP)

form

powder

mp

45-46 °C

storage temp.

2-8°C

SMILES string

[15NH2][13C]#[15N]

InChI

1S/CH2N2/c2-1-3/h2H2/i1+1,2+1,3+1

InChI key

XZMCDFZZKTWFGF-VMIGTVKRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

PRTR

Class II Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

709387-BULK:
709387-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service