708763
[(S)-(−)-2,2′-Bis(di-p-tolylphosphino)-1,1′-binaphthyl]palladium(II) chloride
Synonym(s):
[PdCl2{(S)-4-tolylbinap}]
About This Item
Recommended Products
form
solid
mp
>300 °C
functional group
phosphine
SMILES string
Cl[Pd]Cl.Cc1ccc(cc1)P(c2ccc(C)cc2)c3ccc4ccccc4c3-c5c(ccc6ccccc56)P(c7ccc(C)cc7)c8ccc(C)cc8
InChI
1S/C48H40P2.2ClH.Pd/c1-33-13-23-39(24-14-33)49(40-25-15-34(2)16-26-40)45-31-21-37-9-5-7-11-43(37)47(45)48-44-12-8-6-10-38(44)22-32-46(48)50(41-27-17-35(3)18-28-41)42-29-19-36(4)20-30-42;;;/h5-32H,1-4H3;2*1H;/q;;;+2/p-2
InChI key
ADMKISPNGKUJAU-UHFFFAOYSA-L
Application
- Stereoselective preparation of hydroxymethyldihydronaphthalenone via alkylative ring opening of oxabenzonorbornadiene with (PhMe2SiCH2)2Zn followed by hydrogenation and Tamao-Fleming oxidation
- Asymmetric synthesis of cyclopentylindanes via cyclization of bromo- and trifluoromethanesulfonyloxyphenylheptenylmalonate
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
708763-BULK:
708763-VAR:
708763-100MG:
708763-500MG:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service