Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

703141

Sigma-Aldrich

DABCO®-CuCl complex

97%

Synonym(s):

(1,4-Diazabicyclo[2.2.2]octane)copper(I)chloride complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H12N2 · CuCl
CAS Number:
Molecular Weight:
211.17
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

form

powder

reaction suitability

core: copper
reagent type: catalyst

mp

180-186 °C

SMILES string

Cl[Cu].C1CN2CCN1CC2

InChI

1S/C6H12N2.ClH.Cu/c1-2-8-5-3-7(1)4-6-8;;/h1-6H2;1H;/q;;+1/p-1

InChI key

WHZBKEDMTQJFEI-UHFFFAOYSA-M

Application

Catalyst used for the oxidation of alcohols under mild conditions

Legal Information

DABCO is a registered trademark of Air Products & Chemicals, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

703141-VAR:
703141-BULK:
703141-1G:4548174012581


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mannam, S. et al.
Advanced Synthesis & Catalysis, 2253-2253 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service