Skip to Content
Merck
All Photos(1)

Key Documents

703001

Sigma-Aldrich

3-(Hydroxymethyl)phenylboronic acid pinacol ester

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19BO3
CAS Number:
Molecular Weight:
234.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Quality Level

Assay

96%

form

powder

mp

42-46 °C

storage temp.

2-8°C

SMILES string

CC1(C)OB(OC1(C)C)c2cccc(CO)c2

InChI

1S/C13H19BO3/c1-12(2)13(3,4)17-14(16-12)11-7-5-6-10(8-11)9-15/h5-8,15H,9H2,1-4H3

InChI key

ZEWWJJQAFTXUIS-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ran Gao et al.
British journal of pharmacology, 177(12), 2872-2885 (2020-02-20)
Abdominal aorticaneurysm (AAA) rupture is mainly due to elastic lamina degradation. As a metalloendopeptidase, meprin-α (Mep1A) critically modulates the activity of proteins and inflammatory cytokines in various diseases. Here, we sought to investigate the functional role of Mep1A in AAA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service