Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

698148

Sigma-Aldrich

3-Nitrophenylboronic acid MIDA ester

97%

Synonym(s):

6-Methyl-2-(3-nitrophenyl)-1,3,6,2-dioxazaborocane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11BN2O6
CAS Number:
Molecular Weight:
278.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

211-215 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2cccc(c2)[N+]([O-])=O

InChI

1S/C11H11BN2O6/c1-13-6-10(15)19-12(20-11(16)7-13)8-3-2-4-9(5-8)14(17)18/h2-5H,6-7H2,1H3

InChI key

COEHPTUYNRXODY-UHFFFAOYSA-N

Application

3-Nitrophenylboronic acid MIDA ester is a stable boronic acid surrogate that can be used as a reagent in the Suzuki-Miyaura cross-coupling reactions to prepare useful building blocks via C-C bond formation. 3-allylnitrobenzene is one of the building blocks prepared from this ester by reacting with allyl bromide in the presence of a palladium catalyst.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

698148-BULK:
698148-VAR:
698148-5G:
698148-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MIDA boronate allylation-synthesis of ibuprofen
Phillips D, et al.,
Royal Society of Chemistry Advances, 10(51), 30624-30630 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service