Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

696846

Sigma-Aldrich

3-(Dimethylaminomethyl)-7-azaindole

97%

Synonym(s):

3-(Dimethylaminomethyl)-1H-pyrrolo[2,3-b]pyridine, 7-Azagramine, Dimethyl-(1H-pyrrolo[2,3-b]-3-pyridinylmethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N3
CAS Number:
Molecular Weight:
175.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

144-149 °C

functional group

amine

SMILES string

CN(C)Cc1c[nH]c2ncccc12

InChI

1S/C10H13N3/c1-13(2)7-8-6-12-10-9(8)4-3-5-11-10/h3-6H,7H2,1-2H3,(H,11,12)

InChI key

RFLCFQLBXWLHKX-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

696846-BULK:
696846-1G:
696846-VAR:
696846-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service