Skip to Content
Merck
All Photos(1)

Documents

Safety Information

696609

Sigma-Aldrich

4-Methylthiophene-2-boronic acid pinacol ester

97%

Synonym(s):

2-(4-Methyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene, 4-Methyl-2-thienylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17BO2S
CAS Number:
Molecular Weight:
224.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

liquid

mp

26-31 °C

SMILES string

Cc1csc(c1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C11H17BO2S/c1-8-6-9(15-7-8)12-13-10(2,3)11(4,5)14-12/h6-7H,1-5H3

InChI key

FNRMPXJVCSMVMA-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

696609-VAR:
696609-5G:
696609-BULK:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service