695807
Ethyl 4-(4-methoxyphenyl)-2,4-dioxobutanoate
97%
About This Item
Recommended Products
Assay
97%
form
solid
mp
52-57 °C
SMILES string
CCOC(=O)C(=O)CC(=O)c1ccc(OC)cc1
InChI
1S/C13H14O5/c1-3-18-13(16)12(15)8-11(14)9-4-6-10(17-2)7-5-9/h4-7H,3,8H2,1-2H3
InChI key
JHWYUCIVJQZDEF-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
695807-1G:
695807-BULK:
695807-5G:
695807-VAR:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service