Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

693022

Sigma-Aldrich

(S)-DM-BINAP

Synonym(s):

(S)-(−)-2,2′-Bis[bis(3,5-dimethylphenyl)phosphino]-1,1′-binaphthyl, (S)-(−)-2,2′-Bis[di(3,5-xylyl)phoshino]-1,1′-binaphthyl, (S)-3,5-Xylyl-BINAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H48P2
CAS Number:
Molecular Weight:
734.89
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

solid

optical activity

[α]/D -172°, 20, c = 1 in chloroform

mp

189-193 °C

SMILES string

Cc1cc(C)cc(c1)P(c2cc(C)cc(C)c2)c3ccc4ccccc4c3-c5c(ccc6ccccc56)P(c7cc(C)cc(C)c7)c8cc(C)cc(C)c8

InChI

1S/C52H48P2/c1-33-21-34(2)26-43(25-33)53(44-27-35(3)22-36(4)28-44)49-19-17-41-13-9-11-15-47(41)51(49)52-48-16-12-10-14-42(48)18-20-50(52)54(45-29-37(5)23-38(6)30-45)46-31-39(7)24-40(8)32-46/h9-32H,1-8H3

InChI key

MXGXXBYVDMVJAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Asymmetric hydrogenation of benzophenone
  • Regio- and stereoselective preparation of axially chiral arylnaphthalene derivatives via rhodium-catalyzed [2+2+2] cycloaddition of diynes with naphthalenepropynoic acid derivatives in the presence of chiral biaryl bisphosphine ligands
  • Regiodivergent rhodium-catalyzed [(2+2)+2] carbocyclization of 1,6-enynes with Me propiolates
  • Ligand controlled regioselective and stereoselective desymmetrizing rhodium-catalyzed allylic arylation of meso cyclopentene dicarbonates with arylboronic acids to form regioisomeric arylcyclopentenols
  • Platinum(II) complex-catalyzed enantioselective aldol reaction with ketene silyl acetals in DMF at room temperature
  • Stereoselective preparation of chiral N,O-biaryls via Rh-catalyzed [2+2+2] cycloaddition of conjugate ynamides with diynes

Legal Information

Sold in collaboration with Takasago for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

693022-BULK:
693022-50MG:
693022-VAR:
693022-100MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service