Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

692891

Sigma-Aldrich

5-Bromo-2-(methylthio)pyridine

97%

Synonym(s):

5-Bromo-2-(methylsulfanyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrNS
CAS Number:
Molecular Weight:
204.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

36-41 °C

functional group

bromo
thioether

storage temp.

2-8°C

SMILES string

CSc1ccc(Br)cn1

InChI

1S/C6H6BrNS/c1-9-6-3-2-5(7)4-8-6/h2-4H,1H3

InChI key

CFBYLVDSPYTKPR-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

692891-BULK:
692891-VAR:
692891-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service