Skip to Content
Merck
All Photos(1)

Documents

Safety Information

691216

Sigma-Aldrich

3-(tert-Butoxy)propylamine hydrochloride

97%

Synonym(s):

3-Aminopropyl tert-butyl ether hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H3C)3COCH2CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
167.68
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥96.5% (GC)
96.5-103.5% (with AgNO3, T)
97%

form

powder

storage temp.

2-8°C

SMILES string

Cl.CC(C)(C)OCCCN

InChI

1S/C7H17NO.ClH/c1-7(2,3)9-6-4-5-8;/h4-6,8H2,1-3H3;1H

InChI key

PWSZRKJMCDQAOY-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

691216-BULK:
691216-1G:
691216-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service