690503
2,3-Bis(2,6-diisopropylphenylimino)butane
98%
Synonym(s):
N,N′-(1,2-dimethyl-1,2-ethanediylidene)bis[2,6-bis(1-methylethyl)benzenamine, N,N′-Bis(2,6-diisopropylphenyl)-2,3-butanediimine, Diacetyl-bis(2,6-isopropylphenylimine)
About This Item
Recommended Products
Assay
97.5-102.5% (with HClO4, titration)
98%
form
lumps
reaction suitability
reagent type: ligand
SMILES string
CC(C)c1cccc(C(C)C)c1\N=C(C)\C(C)=N\c2c(cccc2C(C)C)C(C)C
InChI
1S/C28H40N2/c1-17(2)23-13-11-14-24(18(3)4)27(23)29-21(9)22(10)30-28-25(19(5)6)15-12-16-26(28)20(7)8/h11-20H,1-10H3/b29-21+,30-22+
InChI key
YUFQUBWPYIPRHZ-VFIVCBTMSA-N
Application
- Lanthanide(II)-alkali sandwich complexes
- Imido-amido Hf complexes
- Alkali metal compounds
- Tetracoordinated molybdenum(II) complexes
Reactant for:
- Complexation of dimethylmagnesium with α-diimines
- Transfer of tin and germanium atoms from N-heterocyclic stannylenes and germylenes to diazadienes
Hazard Statements
Hazard Classifications
Aquatic Chronic 4
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
690503-VAR:
690503-500MG:
690503-BULK:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service