Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

689211

Sigma-Aldrich

1,4-Butanediol bis(3-aminocrotonate)

Lonza quality, 96.0-100.3 % (w/w) (T)

Synonym(s):

1,4-Butylenglycolbis(3-aminocrotonate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3C(NH2)=CHCO2CH2CH2-]2
CAS Number:
Molecular Weight:
256.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

Lonza quality

form

powder

manufacturer/tradename

Lonza Ltd

concentration

96.0-100.3 % (w/w) (T)

color

white to yellow

mp

146.0-150.0 °C
147-150 °C (lit.)

SMILES string

C\C(N)=C/C(=O)OCCCCOC(=O)\C=C(/C)N

InChI

1S/C12H20N2O4/c1-9(13)7-11(15)17-5-3-4-6-18-12(16)8-10(2)14/h7-8H,3-6,13-14H2,1-2H3/b9-7+,10-8+

InChI key

DFUXQARZEWMFLQ-FIFLTTCUSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

689211-VAR:
689211-BULK:
689211-5KG:
689211-1KG:
689211-20KG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service