688134
4-(4-Chlorophenyl)-2-hydroxythiazole
97%
Synonym(s):
4-(4-Chlorophenyl)-2-thiazolol
About This Item
Assay
97%
form
solid
mp
222-226 °C
SMILES string
Oc1nc(cs1)-c2ccc(Cl)cc2
InChI
1S/C9H6ClNOS/c10-7-3-1-6(2-4-7)8-5-13-9(12)11-8/h1-5H,(H,11,12)
InChI key
XODFWCAVOQHJFC-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service