Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

685852

Sigma-Aldrich

(S)-4-Fluoro-α-methylbenzyl alcohol

97%

Synonym(s):

(S)-1-(4′-Fluorophenyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH(CH3)OH
CAS Number:
Molecular Weight:
140.15
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]20/D -49.0°, c = 1 in chloroform

refractive index

n20/D 1.502

density

1.111 g/mL at 25 °C

functional group

fluoro
hydroxyl

SMILES string

C[C@H](O)c1ccc(F)cc1

InChI

1S/C8H9FO/c1-6(10)7-2-4-8(9)5-3-7/h2-6,10H,1H3/t6-/m0/s1

InChI key

PSDSORRYQPTKSV-LURJTMIESA-N

Application

(S)-4-Fluoro-α-methylbenzyl alcohol can be used as a starting material to synthesize a CCR5 antagonist named MLN1251.[1]Along with its ortho- and non-fluorine substituted analogs, it can be used to study the role of fluorine substitution in chiral discrimination in molecular complexes.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

685852-VAR:
685852-1G:
685852-250MG:
685852-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of fluorine substitution on chiral recognition: interplay of CH? pi, OH? pi and CH? F interactions in gas-phase complexes of 1-aryl-1-ethanol with butan-2-ol
Ciavardini A, et al.
Physical Chemistry Chemical Physics, 15(44), 19360-19370 (2013)
Expedient synthesis of MLN1251, a CCR5 antagonist for treatment of HIV
Ronn M, et al.
Organic Process Research & Development, 11(2), 241-245 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service