Skip to Content
Merck
All Photos(2)

Documents

Safety Information

685399

Sigma-Aldrich

Methyl 1H-pyrrole-3-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO2
CAS Number:
Molecular Weight:
125.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

85-89 °C

SMILES string

COC(=O)c1cc[nH]c1

InChI

1S/C6H7NO2/c1-9-6(8)5-2-3-7-4-5/h2-4,7H,1H3

InChI key

WLBNVSIQCFHAQB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

685399-BULK:
685399-250MG:
685399-VAR:
685399-1G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ahmed M Zheoat et al.
Fitoterapia, 134, 5-13 (2019-01-29)
Hibiscus sabdariffa (Malvaceae) is a plant that is widely recognised for its antihypertensive properties; however the constituent(s) responsible for this biological activity are presently unknown. The aim of this study was to identify the potential compounds that are responsible for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service