Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

684325

Sigma-Aldrich

tert-Butyldimethylsilyl (S)-(+)-glycidyl ether

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20O2Si
CAS Number:
Molecular Weight:
188.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]22/D +6.5°, neat

refractive index

n20/D 1.4320

bp

197-199 °C/760 mmHg

density

0.890 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)[Si](C)(C)OC[C@@H]1CO1

InChI

1S/C9H20O2Si/c1-9(2,3)12(4,5)11-7-8-6-10-8/h8H,6-7H2,1-5H3/t8-/m0/s1

InChI key

YANSSVVGZPNSKD-QMMMGPOBSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.0 °F - closed cup

Flash Point(C)

73.89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

684325-BULK:
684325-VAR:
684325-5ML:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service