683884
2-Cyanophenylboronic acid pinacol ester
97%
Synonym(s):
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
About This Item
Recommended Products
Assay
97%
form
solid
mp
85-90 °C
functional group
nitrile
SMILES string
CC1(C)OB(OC1(C)C)c2ccccc2C#N
InChI
1S/C13H16BNO2/c1-12(2)13(3,4)17-14(16-12)11-8-6-5-7-10(11)9-15/h5-8H,1-4H3
InChI key
SKQNWSBNAIOCOC-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PDSCL
Deleterious substance
JAN Code
683884-VAR:
683884-5G:4548173292496
683884-1G:4548173292489
683884-BULK:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service