Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

682160

Sigma-Aldrich

4-Bromotoluene-d7

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7D7Br
Molecular Weight:
178.08
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

98 atom % D

bp

184 °C (lit.)

mp

26-29 °C (lit.)

density

1.446 g/mL at 25 °C (lit.)

mass shift

M+7

SMILES string

[2H]c1c([2H])c(c([2H])c([2H])c1Br)C([2H])([2H])[2H]

InChI

1S/C7H7Br/c1-6-2-4-7(8)5-3-6/h2-5H,1H3/i1D3,2D,3D,4D,5D

InChI key

ZBTMRBYMKUEVEU-AAYPNNLASA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

682160-VAR:
682160-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ah Reum Lee et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 10(14), e2205913-e2205913 (2023-03-25)
Entosis is a non-apoptotic cell death process that forms characteristic cell-in-cell structures in cancers, killing invading cells. Intracellular Ca2+ dynamics are essential for cellular processes, including actomyosin contractility, migration, and autophagy. However, the significance of Ca2+ and Ca2+ channels participating
Sangwoo Shim et al.
Proceedings of the National Academy of Sciences of the United States of America, 120(23), e2220037120-e2220037120 (2023-05-30)
The balance between neural stem cell proliferation and neuronal differentiation is paramount for the appropriate development of the nervous system. Sonic hedgehog (Shh) is known to sequentially promote cell proliferation and specification of neuronal phenotypes, but the signaling mechanisms responsible
Mustafa Yalcinkaya et al.
Journal of lipid research, 65(4), 100534-100534 (2024-03-25)
The deposition of cholesterol-rich lipoproteins in the arterial wall triggers macrophage inflammatory responses, which promote atherosclerosis. The NLRP3 inflammasome aggravates atherosclerosis; however, cellular mechanisms connecting macrophage cholesterol accumulation to inflammasome activation are poorly understood. We investigated the mechanisms of NLRP3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service