Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

681911

Sigma-Aldrich

N-Boc-indoline-7-carboxaldehyde

97%

Synonym(s):

1-(tert-Butoxycarbonyl)indoline-7-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO3
CAS Number:
Molecular Weight:
247.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

86-90 °C

functional group

aldehyde

SMILES string

[H]C(=O)c1cccc2CCN(C(=O)OC(C)(C)C)c12

InChI

1S/C14H17NO3/c1-14(2,3)18-13(17)15-8-7-10-5-4-6-11(9-16)12(10)15/h4-6,9H,7-8H2,1-3H3

InChI key

KUYIMEZLCAWOMI-UHFFFAOYSA-N

Application

Useful bi-functional starter in indoline chemistry and natural product chemistry

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

681911-1G:
681911-VAR:
681911-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Syntheses, 9, 124-124 (1998)
Organic Syntheses, 73, 85-85 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service