Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

678546

Sigma-Aldrich

(−)-Ipc2B(allyl)borane solution

1 M in dioxane

Synonym(s):

(−)-B-Allyldiisopinocampheylborane solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H17)2BCH2CH=CH2
CAS Number:
Molecular Weight:
326.37
UNSPSC Code:
12352000

optical activity

[α]20/D −35 to ±5°, c = 1

concentration

1 M in dioxane

bp

100-102 °C

density

0.964 g/mL at 25 °C

SMILES string

C[C@H]1[C@@H](C[C@@H]2C[C@H]1C2(C)C)B(CC=C)[C@@H]3C[C@@H]4C[C@H]([C@H]3C)C4(C)C

InChI

1S/C23H39B/c1-8-9-24(20-12-16-10-18(14(20)2)22(16,4)5)21-13-17-11-19(15(21)3)23(17,6)7/h8,14-21H,1,9-13H2,2-7H3/t14-,15-,16+,17+,18-,19-,20-,21-/m1/s1

InChI key

ZIXZBDJFGUIKJS-RLEROFIGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Material may become hazy upon cooler storage, this will not affect use

Application

Convenient, salt-free, stable solution of a chiral allyl borane for asymmetric allylation of aldehydes leading to chiral homoallylic alcohols.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

FSL

Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

678546-5ML:4548173240121
678546-BULK:
678546-VAR:
678546-25ML:4548173240114


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 35, 23-23 (2002)
Toward the total synthesis of spirastrellolide A. Part 1: Strategic considerations and preparation of the southern domain.
Alois Fürstner et al.
Angewandte Chemie (International ed. in English), 45(33), 5506-5510 (2006-07-11)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service