Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

677884

Sigma-Aldrich

Methyl 5-methoxypyridine-3-carboxylate

90%

Synonym(s):

Methyl 5-methoxy-3-pyridinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC5H3NCOOCH3
CAS Number:
Molecular Weight:
167.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

90%

form

solid

mp

54-59 °C

SMILES string

COC(=O)c1cncc(OC)c1

InChI

1S/C8H9NO3/c1-11-7-3-6(4-9-5-7)8(10)12-2/h3-5H,1-2H3

InChI key

JIRJAOSTLIQOCY-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

677884-1G:
677884-250MG:
677884-VAR:
677884-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service