Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

671614

Sigma-Aldrich

4-Ethyl-4-methylmorpholinium methyl carbonate solution

~50% in methanol: water (2:3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO4
CAS Number:
Molecular Weight:
205.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

concentration

~50% in methanol: water (2:3)

refractive index

n20/D 1.408

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤10 mg/kg
fluoride (F-): ≤10 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

SMILES string

COC([O-])=O.CC[N+]1(C)CCOCC1

InChI

1S/C7H16NO.C2H4O3/c1-3-8(2)4-6-9-7-5-8;1-5-2(3)4/h3-7H2,1-2H3;1H3,(H,3,4)/q+1;/p-1

InChI key

JWKQGIUVWAJWAE-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Eyes, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

121.1 °F

Flash Point(C)

49.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

671614-100G:4548173242361
671614-BULK:
671614-VAR:
671614-10G:4548173242378


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

CBILS© offers a simple, elegant synthetic method for the production of ionic liquids, developed by the Austrian company proionic.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service