Skip to Content
Merck
All Photos(1)

Key Documents

670928

(−)-2,3-Bis[(2R,5R)-2,5-dimethylphospholano]maleic anhydride(2,5-norbornadiene)rhodium(I) tetrafluoroborate

Synonym(s):

2,3-Bis[(2R,5R)-2,5-dimethyl-phospholanyl]maleic anhydride(2,5-norbornadiene)rhodium(I) tetrafluoroborate, catASium® M(R)RhNor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32BF4O3P2Rh
CAS Number:
Molecular Weight:
608.16
Pricing and availability is not currently available.

storage temp.

2-8°C

General description

Sold in collaboration with Solvias AG

Legal Information

US20050209455
catASium is a registered trademark of Umicore AG & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service