Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

66947

Sigma-Aldrich

(S)-Boc-3-(2-naphthyl)-β-Ala-OH

≥98.0% (HPLC)

Synonym(s):

(S)-3-(Boc-amino)-3-(2-naphthyl)propionic acid, D-Boc-3-(2-naphthyl)-β-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO4
CAS Number:
Molecular Weight:
315.36
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CC(O)=O)c1ccc2ccccc2c1

InChI

1S/C18H21NO4/c1-18(2,3)23-17(22)19-15(11-16(20)21)14-9-8-12-6-4-5-7-13(12)10-14/h4-10,15H,11H2,1-3H3,(H,19,22)(H,20,21)/t15-/m0/s1

InChI key

NCQJBPXXRXOIJD-HNNXBMFYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

66947-VAR-F:
66947-BULK-F:
66947-500MG-F:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service