Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

668982

Sigma-Aldrich

2,2′-Bis(di-p-tolylphosphino)-1,1′-binaphthyl

97%

Synonym(s):

rac-Tol-BINAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H40P2
CAS Number:
Molecular Weight:
678.78
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

reaction suitability

reagent type: ligand

mp

267-271 °C

functional group

phosphine

SMILES string

Cc1ccc(cc1)P(c2ccc(C)cc2)c3ccc4ccccc4c3-c5c(ccc6ccccc56)P(c7ccc(C)cc7)c8ccc(C)cc8

InChI

1S/C48H40P2/c1-33-13-23-39(24-14-33)49(40-25-15-34(2)16-26-40)45-31-21-37-9-5-7-11-43(37)47(45)48-44-12-8-6-10-38(44)22-32-46(48)50(41-27-17-35(3)18-28-41)42-29-19-36(4)20-30-42/h5-32H,1-4H3

InChI key

IOPQYDKQISFMJI-UHFFFAOYSA-N

Application

Used in:
  • Rhodium-catalyzed 2+2+2 cycloaddition reactions
  • Decarboxylative coupling of carboxylic acids using copper-palladium-phenanthroline-Tol-BINAP catalyst system
  • Palladium-catalyzed coupling of aromatic iodides or bromides and tin-thiolates under microwave conditions
  • Preparation of methylenesultams via intramolecular Heck cyclization of bromovinylsulfonamides and subsequent Michael addition reaction of sultams with thiols
  • Regioselective intermolecular cyclotrimerization of alkynes in complex with cationic rhodium(I)
  • Palladium-catalyzed amination reactions
  • Palladium-catalyzed ether formation from aryl halides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

668982-1G:
668982-BULK:
668982-VAR:
668982-250MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service